siRNA Details
| Gene Symbol: Adprm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025510.3 | NP_079786.2 | 3 | 287 | ||
| GenBank mRNA | AK009142.1 | - | 3 | 265 | ||
| AK010520.1 | - | 2 | 247 | |||
| AK145941.1 | - | 3 | 230 | |||
| BY118851.1 | - | 3 | 287 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adprm| Gene Symbol: Adprm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025510.3 | NP_079786.2 | 3 | 287 | ||
| GenBank mRNA | AK009142.1 | - | 3 | 265 | ||
| AK010520.1 | - | 2 | 247 | |||
| AK145941.1 | - | 3 | 230 | |||
| BY118851.1 | - | 3 | 287 | |||