siRNA Details
| Gene Symbol: Adprm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025510.3 | NP_079786.2 | 3 | 455 | ||
| GenBank mRNA | AK009142.1 | - | 3 | 433 | ||
| AK145941.1 | - | 3 | 398 | |||
| BC004029.1 | - | 3 | 407 | 3495093 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adprm| Gene Symbol: Adprm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025510.3 | NP_079786.2 | 3 | 455 | ||
| GenBank mRNA | AK009142.1 | - | 3 | 433 | ||
| AK145941.1 | - | 3 | 398 | |||
| BC004029.1 | - | 3 | 407 | 3495093 | ||