siRNA Details
| Gene Symbol: Ndufc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025523.1 | NP_079799.1 | 2 | 130 | ||
| GenBank mRNA | AK003365.1 | - | 2 | 116 | ||
| AK009393.1 | - | 2 | 130 | |||
| AK018705.1 | - | 2 | 114 | |||
| AK028011.1 | - | 114 | ||||
| BC092141.1 | - | 2 | 122 | 6814605 | ||

Ndufc1
Genomic Map