siRNA Details
| Gene Symbol: Ccdc82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025534.2 | NP_079810.2 | 3 | 709 | ||
| GenBank mRNA | AK009786.1 | - | 2 | 609 | ||
| AK141751.1 | - | 3 | 707 | |||
| AK166406.1 | - | 2 | 649 | |||
| BC057326.1 | - | 3 | 709 | 6826080 | ||
| BC098496.1 | - | 2 | 636 | 30089818 | ||

Ccdc82
Genomic Map