siRNA Details
| Gene Symbol: Alkbh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025538.3 | NP_079814.1 | 3 | 437 | ||
| NM_027372.1 | NP_081648.1 | 2 | 263 | |||
| GenBank mRNA | AK007380.1 | - | 3 | 415 | ||
| AK009812.1 | - | 3 | 424 | |||
| AK010930.1 | - | 2 | 264 | |||
| BC029677.1 | - | 3 | 404 | 5364485 | ||

Alkbh7
Genomic Map