siRNA Details
| Gene Symbol: Sac3d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133678.3 | NP_598439.3 | 1 | 443 | ||
| GenBank mRNA | AJ131957.2 | - | 1 | 443 | ||
| AK011298.1 | - | 1 | 357 | |||
| AK014138.1 | - | 1 | 397 | |||
| AK075930.1 | - | 1 | 403 | |||
| AK139556.1 | - | 1 | 405 | |||
| BC144812.1 | - | 1 | 507 | 9053360 | ||
| BC145789.1 | - | 1 | 507 | 40131092 | ||

Sac3d1
Genomic Map