siRNA Details
| Gene Symbol: Ndufa12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025551.3 | NP_079827.2 | 4 | 353 | ||
| GenBank mRNA | AK003131.1 | - | 4 | 326 | ||
| AK003322.1 | - | 4 | 326 | |||
| AK008876.1 | - | 4 | 326 | |||
| AK010460.1 | - | 4 | 354 | |||
| AK032774.1 | - | 4 | 327 | |||
| AK134765.1 | - | 4 | 326 | |||
| AK153240.1 | - | 4 | 326 | |||
| AK167943.1 | - | 4 | 319 | |||
| BC004633.1 | - | 1783 | ||||
| BC086922.1 | - | 4 | 358 | 6751147 | ||

Ndufa12
Genomic Map