siRNA Details
| Gene Symbol: Oxld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025560.2 | NP_079836.1 | 1, 2 | 249 | ||
| GenBank mRNA | AK007842.1 | - | 1, 2 | 250 | ||
| AK021150.1 | - | 1, 2 | 237 | |||
| BC147291.1 | - | 1, 2 | 242 | 9056342 | ||
| BC147292.1 | - | 1, 2 | 242 | 9056341 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BC051593.1 | - | 578 | Chr. - - [-] on Build | ||

Oxld1
Genomic Map