siRNA Details
| Gene Symbol: Uggt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081252.2 | NP_001074721.2 | 15 | 1682 | ||
| GenBank mRNA | AK038817.1 | - | 15 | 1678 | ||
| AK046834.1 | - | 2 | 161 | |||
| BC110672.1 | - | 1 | 63 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uggt2| Gene Symbol: Uggt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081252.2 | NP_001074721.2 | 15 | 1682 | ||
| GenBank mRNA | AK038817.1 | - | 15 | 1678 | ||
| AK046834.1 | - | 2 | 161 | |||
| BC110672.1 | - | 1 | 63 | |||