siRNA Details
| Gene Symbol: Tnfaip8l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025566.3 | NP_079842.2 | 2 | 232 | ||
| GenBank mRNA | AK004949.1 | - | 2 | 164 | ||
| AK011245.1 | - | 2 | 194 | |||
| AK090348.1 | - | 2 | 234 | |||
| AK136962.1 | - | 2 | 194 | |||
| AK150678.1 | - | 2 | 223 | |||
| AK153177.1 | - | 2 | 218 | |||
| BC032199.1 | - | 2 | 161 | 5324284 | ||
| BY733616.1 | - | 2 | 232 | |||

Tnfaip8l1
Genomic Map