siRNA Details
| Gene Symbol: Ptpmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025576.2 | NP_079852.1 | 4 | 824 | ||
| GenBank mRNA | AK003210.1 | - | 4 | 528 | ||
| AK012674.1 | - | 4 | 814 | |||
| AK014691.1 | - | 4 | 818 | |||
| AK151972.1 | - | 4 | 825 | |||
| AK168416.1 | - | 690 | ||||
| AK219167.1 | - | 1 | 61 | |||
| BC026750.1 | - | 4 | 803 | 5135841 | ||

Ptpmt1
Genomic Map