siRNA Details
| Gene Symbol: 1700029F12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080777.2 | NP_001074246.1 | 1 | 163 | ||
| NM_025585.3 | NP_079861.1 | 1 | 163 | |||
| GenBank mRNA | AK006036.1 | - | 1 | 151 | ||
| AK006488.1 | - | 1 | 131 | |||
| BC049576.1 | - | 1 | 137 | 6772947 | ||

1700029F12Rik
Genomic Map