siRNA Details
Gene Symbol: 1700029F12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080777.2 | NP_001074246.1 | 2 | 254 | ||
NM_001284205.1 | NP_001271134.1 | 2 | 212 | |||
NM_025585.3 | NP_079861.1 | 2 | 254 | |||
GenBank mRNA | AK006036.1 | - | 2 | 242 | ||
AK006488.1 | - | 2 | 222 | |||
BC049576.1 | - | 2 | 228 | 6772947 | ||
BU962717.1 | - | 2 | 215 | |||
CF106098.1 | - | 1 | 64 |
