siRNA Details
Gene Symbol: Nova1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021361.1 | NP_067336.1 | 2 | 199 | ||
GenBank mRNA | AF232828.1 | - | 2 | 133 | ||
BM942596.1 | - | 2 | 199 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001100541, XM_006240070, XM_006240069, AY262017.
Panther Classification:
Gene Ontology Categories:
