siRNA Details
| Gene Symbol: Nova1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021361.1 | NP_067336.1 | 2 | 277 | ||
| GenBank mRNA | AF232828.1 | - | 2 | 211 | ||
| AK090295.1 | - | 2 | 566 | |||
| BM942596.1 | - | 2 | 277 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nova1| Gene Symbol: Nova1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021361.1 | NP_067336.1 | 2 | 277 | ||
| GenBank mRNA | AF232828.1 | - | 2 | 211 | ||
| AK090295.1 | - | 2 | 566 | |||
| BM942596.1 | - | 2 | 277 | |||