siRNA Details
| Gene Symbol: Cmss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025599.3 | NP_079875.2 | 10 | 917 | ||
| GenBank mRNA | AK012168.1 | - | 10 | 898 | ||
| AK019979.1 | - | 10 | 893 | |||
| BC038891.1 | - | 10 | 804 | 3371329 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cmss1| Gene Symbol: Cmss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025599.3 | NP_079875.2 | 10 | 917 | ||
| GenBank mRNA | AK012168.1 | - | 10 | 898 | ||
| AK019979.1 | - | 10 | 893 | |||
| BC038891.1 | - | 10 | 804 | 3371329 | ||