siRNA Details
| Gene Symbol: Rnf181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025607.3 | NP_079883.3 | 1 | 69 | ||
| GenBank mRNA | AK010854.1 | - | 1 | 82 | ||
| AK159788.1 | - | 1 | 82 | |||
| AK166904.1 | - | 1 | 81 | |||
| AK166995.1 | - | 1 | 82 | |||
| AK167396.1 | - | 1 | 82 | |||
| AK167633.1 | - | 1 | 82 | |||
| AK167935.1 | - | 1 | 82 | |||
| AK168670.1 | - | 1 | 83 | |||
| BC005559.1 | - | 1 | 93 | |||
| BC083119.1 | - | 1 | 70 | 5374213 | ||

Rnf181
Genomic Map