siRNA Details
| Gene Symbol: Clec2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101507.1 | NP_001094977.1 | 2 | 382 | ||
| GenBank mRNA | CJ059176.1 | - | 2 | 173 | ||
| CJ060465.1 | - | 2 | 384 | |||
| CJ060631.1 | - | 2 | 384 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clec2l| Gene Symbol: Clec2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101507.1 | NP_001094977.1 | 2 | 382 | ||
| GenBank mRNA | CJ059176.1 | - | 2 | 173 | ||
| CJ060465.1 | - | 2 | 384 | |||
| CJ060631.1 | - | 2 | 384 | |||