siRNA Details
Gene Symbol: Rwdd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025614.3 | NP_079890.1 | 1 | 193 | ||
GenBank mRNA | AF503942.1 | - | 1 | 85 | ||
AK002967.1 | - | 607 | ||||
AK003262.2 | - | 1 | 147 | |||
AK005278.1 | - | 1 | 105 | |||
AK009510.1 | - | 1 | 62 | |||
AK011277.1 | - | 1 | 193 | |||
AK012504.1 | - | 1 | 164 | |||
BC062136.1 | - | 1 | 75 | 6508909 |
