siRNA Details
| Gene Symbol: Vmn2r28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081405.1 | NP_001074874.1 | 3 | 600 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn2r28| Gene Symbol: Vmn2r28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081405.1 | NP_001074874.1 | 3 | 600 | ||
Set Membership: |
|