siRNA Details
| Gene Symbol: Adamtsl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285435.1 | NP_001272364.1 | 4, 5 | 454 | ||
| GenBank mRNA | AK008355.1 | - | 4, 5 | 419 | ||
| AK008364.1 | - | 4, 5 | 419 | |||
| AK008440.1 | - | 4, 5 | 418 | |||
| AK029081.1 | - | 4, 5 | 395 | |||
| BC034843.1 | - | 2, 3 | 307 | |||

Adamtsl5
Genomic Map