siRNA Details
| Gene Symbol: Sppl2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023220.2 | NP_075709.2 | 13 | 1530 | ||
| GenBank mRNA | AB041547.1 | - | 13 | 1530 | ||
| AK155011.1 | - | 13 | 1329 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sppl2a| Gene Symbol: Sppl2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023220.2 | NP_075709.2 | 13 | 1530 | ||
| GenBank mRNA | AB041547.1 | - | 13 | 1530 | ||
| AK155011.1 | - | 13 | 1329 | |||