siRNA Details
| Gene Symbol: Mthfd2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026788.1 | NP_081064.1 | 4 | 565 | ||
| GenBank mRNA | AK003824.1 | - | 2 | 132 | ||
| BU610697.1 | - | 3 | 246 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mthfd2l| Gene Symbol: Mthfd2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026788.1 | NP_081064.1 | 4 | 565 | ||
| GenBank mRNA | AK003824.1 | - | 2 | 132 | ||
| BU610697.1 | - | 3 | 246 | |||