siRNA Details
| Gene Symbol: Rwdd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025637.3 | NP_079913.2 | 2 | 189 | ||
| GenBank mRNA | AK011007.1 | - | 2 | 189 | ||
| AK011008.1 | - | 2 | 189 | |||
| AK050254.1 | - | 1 | 56 | |||
| BC128288.1 | - | 2 | 188 | |||
| BC150838.1 | - | 2 | 189 | 9087749 | ||
| BY710389.1 | - | 2 | 189 | |||

Rwdd3
Genomic Map