siRNA Details
| Gene Symbol: Mis18a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025642.1 | NP_079918.1 | 1 | 254 | ||
| GenBank mRNA | AK011719.1 | - | 1 | 105 | ||
| AK012533.1 | - | 1 | 255 | |||
| AK014084.1 | - | 1 | 223 | |||
| AK163310.1 | - | 1 | 288 | |||
| BC079900.1 | - | 1 | 266 | 30602551 | ||

Mis18a
Genomic Map