siRNA Details
Gene Symbol: Fastk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023229.2 | NP_075718.2 | 2 | 187 | ||
GenBank mRNA | AF168682.1 | - | 1 | 82 | ||
AK002552.1 | - | 2 | 139 | |||
AK052667.1 | - | 2 | 132 | |||
AK139374.1 | - | 1 | 905 | |||
AK151859.1 | - | 2 | 139 | |||
AK152369.1 | - | 2 | 188 | |||
AK159751.1 | - | 2 | 139 | |||
AK164317.1 | - | 146 | ||||
AK166508.1 | - | 903 | ||||
AK167346.1 | - | 1 | 899 | |||
AK177318.1 | - | 1 | 28 | |||
BC013547.1 | - | 2 | 104 | 4218374 |
