siRNA Details
| Gene Symbol: Diablo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023232.3 | NP_075721.3 | 6 | 744 | ||
| GenBank mRNA | AF203914.1 | - | 6 | 710 | ||
| AK002887.1 | - | 6 | 712 | |||
| AK012760.1 | - | 6 | 742 | |||
| AK077198.1 | - | 6 | 725 | |||
| AK082903.1 | - | 5 | 835 | |||
| AK133194.1 | - | 724 | ||||
| BC054109.1 | - | 6 | 738 | 30013833 | ||

Diablo
Genomic Map