siRNA Details
| Gene Symbol: Gtf3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025652.3 | NP_079928.2 | 3 | 436 | ||
| GenBank mRNA | AF391799.1 | - | 3 | 420 | ||
| AK008247.2 | - | 3 | 403 | |||
| AK011845.1 | - | 3 | 385 | |||
| AK012797.1 | - | 3 | 383 | |||
| AK017234.1 | - | 3 | 330 | |||
| AK143627.1 | - | 3 | 388 | |||
| BC032292.1 | - | 2 | 91 | 5374268 | ||
| BC057966.1 | - | 2 | 648 | |||

Gtf3a
Genomic Map