siRNA Details
| Gene Symbol: Gtf3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025652.3 | NP_079928.2 | 6, 7 | 768 | ||
| GenBank mRNA | AF391799.1 | - | 6, 7 | 752 | ||
| AK008247.2 | - | 6, 7 | 736 | |||
| AK012797.1 | - | 6, 7 | 715 | |||
| AK017234.1 | - | 6, 7 | 662 | |||
| AK143627.1 | - | 6, 7 | 720 | |||
| BC064891.1 | - | 4, 5 | 439 | |||

Gtf3a
Genomic Map