siRNA Details
| Gene Symbol: Gemin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025656.4 | NP_079932.2 | 6, 7 | 613 | ||
| GenBank mRNA | AK005928.1 | - | 6, 7 | 615 | ||
| AK007515.1 | - | 6, 7 | 614 | |||
| AK013414.1 | - | 6, 7 | 570 | |||
| BC053424.1 | - | 6, 7 | 1042 | 6336522 | ||

Gemin2
Genomic Map