siRNA Details
| Gene Symbol: Snx9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025664.5 | NP_079940.2 | 17 | 1879 | ||
| GenBank mRNA | AK004449.1 | - | 250 | |||
| AK012520.1 | - | 12 | 1171 | |||
| AK139802.1 | - | 17 | 1913 | |||
| AK149739.1 | - | 17 | 1892 | |||
| AK149849.1 | - | 17 | 1933 | |||
| AK155830.1 | - | 1914 | ||||
| AK157517.1 | - | 17 | 1895 | |||
| AK170339.1 | - | 1895 | ||||
| AK189192.1 | - | 3 | 214 | |||
| BC014814.1 | - | 17 | 1879 | 3257422 | ||

Snx9
Genomic Map