siRNA Details
| Gene Symbol: Snrnp27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025665.2 | NP_079941.1 | 2 | 69 | ||
| GenBank mRNA | AK011937.1 | - | 2 | 70 | ||
| AK045231.1 | - | 2 | 82 | |||
| AK160190.1 | - | 2 | 79 | |||
| BC027564.1 | - | 2 | 70 | 3468252 | ||

Snrnp27
Genomic Map