siRNA Details
| Gene Symbol: Spcs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025668.3 | NP_079944.1 | 3 | 303 | ||
| GenBank mRNA | AK014310.1 | - | 3 | 317 | ||
| AK017504.1 | - | 3 | 308 | |||
| AK077510.1 | - | 3 | 313 | |||
| AK134578.1 | - | 3 | 304 | |||
| AK168173.1 | - | 3 | 308 | |||
| BC010547.1 | - | 2 | 121 | |||
| BC050150.1 | - | 3 | 313 | 30026746 | ||

Spcs2
Genomic Map