siRNA Details
| Gene Symbol: Tsen15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025677.3 | NP_079953.2 | 2 | 287 | ||
| GenBank mRNA | AK004261.1 | - | 2 | 223 | ||
| AK011608.1 | - | 2 | 244 | |||
| AK017650.1 | - | 2 | 288 | |||
| AK169429.1 | - | 2 | 2741 | |||
| BC024342.1 | - | 2 | 217 | 5002659 | ||
| BC145701.1 | - | 2 | 254 | 40131004 | ||
| BY065176.1 | - | 2 | 239 | |||

Tsen15
Genomic Map