siRNA Details
| Gene Symbol: Tsen15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025677.3 | NP_079953.2 | 4 | 490 | ||
| GenBank mRNA | AK004261.1 | - | 4 | 426 | ||
| AK011608.1 | - | 4 | 447 | |||
| AK017650.1 | - | 4 | 491 | |||
| AK169429.1 | - | 3 | 3245 | |||
| BC024342.1 | - | 4 | 420 | 5002659 | ||
| BC145701.1 | - | 4 | 457 | 40131004 | ||

Tsen15
Genomic Map