siRNA Details
| Gene Symbol: Spryd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025697.3 | NP_079973.1 | 4 | 725 | ||
| GenBank mRNA | AK018044.1 | - | 4 | 603 | ||
| AK018153.1 | - | 4 | 726 | |||
| AK018290.1 | - | 4 | 575 | |||
| AK137685.1 | - | 574 | ||||
| AK167306.1 | - | 4 | 567 | |||
| AK169059.1 | - | 4 | 574 | |||
| BC037015.1 | - | 4 | 559 | 5344409 | ||

Spryd7
Genomic Map