siRNA Details
| Gene Symbol: Rbm24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081425.1 | NP_001074894.1 | 4 | 1176 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ159111.1 | - | 344 | Chr. - - [-] on Build | ||

Rbm24
Genomic Map