siRNA Details
| Gene Symbol: Zfp932 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145563.2 | NP_663538.2 | 1 | 79 | ||
| GenBank mRNA | AK009062.1 | - | 1 | 95 | ||
| BC012405.1 | - | 1 | 65 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp932| Gene Symbol: Zfp932 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145563.2 | NP_663538.2 | 1 | 79 | ||
| GenBank mRNA | AK009062.1 | - | 1 | 95 | ||
| BC012405.1 | - | 1 | 65 | |||