siRNA Details
| Gene Symbol: Gapvd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025709.2 | NP_079985.2 | 4 | 1514 | ||
| GenBank mRNA | AK129381.1 | - | 3 | 1109 | ||
| AK134776.1 | - | 1 | 423 | |||
| AK150026.1 | - | 3 | 1123 | |||
| AK165047.1 | - | 3 | 1126 | |||
| BC031478.1 | - | 1 | 559 | |||
| BC032088.1 | - | 1 | 559 | |||
| BC048847.1 | - | 3 | 1080 | |||
| BC060123.1 | - | 4 | 1514 | 6408778 | ||
| EF155419.1 | - | 2 | 932 | |||

Gapvd1
Genomic Map