siRNA Details
| Gene Symbol: Snx31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025712.4 | NP_079988.3 | 5, 6 | 492 | ||
| GenBank mRNA | AK014536.1 | - | 5, 6 | 494 | ||
| AK034196.1 | - | 5, 6 | 494 | |||
| AK035226.1 | - | 5, 6 | 493 | |||
| BC061007.1 | - | 5, 6 | 488 | 30314775 | ||

Snx31
Genomic Map