siRNA Details
Gene Symbol: Chmp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025783.3 | NP_080059.2 | 2 | 221 | ||
GenBank mRNA | AK009414.1 | - | 2 | 95 | ||
AK014818.1 | - | 2 | 159 | |||
AK016677.1 | - | 2 | 145 | |||
AK018611.1 | - | 2 | 176 | |||
AK083562.1 | - | 2 | 222 | |||
BC027441.1 | - | 2 | 85 | |||
BC039759.1 | - | 2 | 96 | |||
BC049964.1 | - | 2 | 142 | 3154282 |
