siRNA Details
Gene Symbol: Sbds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023248.1 | NP_075737.1 | 3 | 436 | ||
GenBank mRNA | AK005079.1 | - | 3 | 413 | ||
AK014648.1 | - | 3 | 441 | |||
AK019228.1 | - | 3 | 423 | |||
AK168847.1 | - | 3 | 423 | |||
BC003849.1 | - | 3 | 436 | 3599733 |
