siRNA Details
Gene Symbol: 9930111J21Rik1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114679.1 | NP_001108151.1 | 2458 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 9930111J21Rik1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114679.1 | NP_001108151.1 | 2458 |