siRNA Details
| Gene Symbol: Spag16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025728.4 | NP_080004.1 | 5 | 660 | ||
| NM_029160.3 | NP_083436.2 | 14 | 1501 | |||
| GenBank mRNA | AF490391.1 | - | 14 | 1483 | ||
| AK014865.1 | - | 5 | 655 | |||
| AK016618.1 | - | 5 | 647 | |||

Spag16
Genomic Map