siRNA Details
| Gene Symbol: Spag16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025728.4 | NP_080004.1 | 8 | 978 | ||
| NM_029160.3 | NP_083436.2 | 17 | 1819 | |||
| GenBank mRNA | AF490391.1 | - | 17 | 1801 | ||
| AK014865.1 | - | 8 | 973 | |||
| AK016618.1 | - | 8 | 965 | |||

Spag16
Genomic Map