siRNA Details
| Gene Symbol: Gm8633 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BQ828582.1 | - | 3, 4 | 182 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm8633| Gene Symbol: Gm8633 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BQ828582.1 | - | 3, 4 | 182 | ||