siRNA Details
| Gene Symbol: Erich2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025744.2 | NP_080020.2 | 10 | 1259 | ||
| GenBank mRNA | AK006384.1 | - | 10 | 587 | ||
| AK017113.1 | - | 10 | 599 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Erich2| Gene Symbol: Erich2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025744.2 | NP_080020.2 | 10 | 1259 | ||
| GenBank mRNA | AK006384.1 | - | 10 | 587 | ||
| AK017113.1 | - | 10 | 599 | |||