siRNA Details
| Gene Symbol: 4933411K16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025752.2 | NP_080028.1 | 1 | 962 | ||
| GenBank mRNA | AK006374.1 | - | 1 | 939 | ||
| AK016775.1 | - | 1 | 943 | |||
| BC049703.1 | - | 1 | 939 | 6707366 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933411K16Rik| Gene Symbol: 4933411K16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025752.2 | NP_080028.1 | 1 | 962 | ||
| GenBank mRNA | AK006374.1 | - | 1 | 939 | ||
| AK016775.1 | - | 1 | 943 | |||
| BC049703.1 | - | 1 | 939 | 6707366 | ||