siRNA Details
| Gene Symbol: Piezo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039485.4 | NP_001034574.4 | 27 | 4930 | ||
| GenBank mRNA | AK087529.1 | - | 26 | 4161 | ||
| HQ215521.1 | - | 27 | 4016 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Piezo2| Gene Symbol: Piezo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039485.4 | NP_001034574.4 | 27 | 4930 | ||
| GenBank mRNA | AK087529.1 | - | 26 | 4161 | ||
| HQ215521.1 | - | 27 | 4016 | |||