siRNA Details
| Gene Symbol: Ube2w | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025773.3 | NP_080049.2 | 4 | 359 | ||
| GenBank mRNA | AK015348.1 | - | 4 | 313 | ||
| AK042870.1 | - | 4 | 309 | |||
| AK044463.1 | - | 4 | 311 | |||
| AK046086.1 | - | 4 | 313 | |||
| AK050679.1 | - | 4 | 311 | |||
| AK077997.1 | - | 4 | 284 | |||
| AK082233.1 | - | 4 | 308 | |||
| BC020124.1 | - | 4 | 311 | 4012785 | ||
| BY728312.1 | - | 4 | 359 | |||
| DV044691.1 | - | 2 | 205 | |||

Ube2w
Genomic Map